Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific Supplies UniStore™ Amber Reagent Bottles, Narrow Mouth, HDPE

Shop All United Scientific Supplies, Inc Products


Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Description
- United Scientific™ Narrow Mouth, HDPE, reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant.
- The amber bottles are designed to reduce UV light transmission to protect light-sensitive contents and conform to USP requirements for storage of light sensitive materials.
- Excellent chemical resistance to most acids, bases, and alcohols.
- Polypropylene caps are included.
- Protect light-sensitive materials during shipping and storage.
- Meet USP requirements for storage of light-sensitive materials.
- Manufactured from USP Class VI resin that is FDA-21CFR compliant for food contact.
- Free of heavy metals, with Leakproof closure with integrated seal ring.
- Precision molded for consistent wall thickness and smooth interior and exterior surfaces.
Specifications
Specifications
| Capacity (Metric) | 60 mL |
| Capacity (English) | 2 oz. |
| Color | Amber |
| Material | HDPE |
| Packaging | Pack |
| Quantity | 72 Pk. |
| Sterility | Non-sterile |
| Product Line | UniStore |
| Closure Included | Yes |
| Disposable | Reusable |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction