Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ UniStore™ Reagent Bottles, Narrow Mouth, PP, Bulk Packed
SDPSCI_ED

Catalog No. S43672 Shop All United Scientific Supplies, Inc Products
Click to view available options
Capacity (Metric):
15 mL
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL

Catalog No. S43672

Supplier: United Scientific™ 33302BULK

Only null left
Add to Cart
Add to Cart

Narrow Mouth Polypropylene Reagent Bottles are rigid, translucent and have fine contact clarity.

UniStore™ Narrow Mouth, PP, reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant.

  • Excellent chemical resistance to most acids, bases, and alcohols.
  • Polypropylene caps are included.
  • Manufactured from USP Class VI resin that is FDA-21CFR compliant for food contact and free of heavy metals.
  • Leakproof closure with integrated seal ring.
  • Precision molded for consistent wall thickness with smooth interior and exterior surfaces.
  • Certificates of Analysis for leakproof test details are available upon request.

Specifications

Capacity (Metric) 125 mL
Capacity (English) 4 oz.
Color Translucent
Diameter (Metric) Inner Neck 18 mm
Material Polypropylene
Packaging Bulk
Quantity 500 Pk.
Sterility Non-sterile
Product Line UniStore
Closure Included No
Graduated No
Mouth Narrow
Height (Metric) 101 mm
Product Type Reagent Bottle
Height (English) 4 in.
Closure Material Polypropylene
Closure Size 24 mm
Diameter (Metric) 18 mm
Through the Closure Dispensing No
Autoclavable Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.