Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NARS2 Antibody (OTI10C10), PE, Novus Biologicals™
SDP

Catalog No. NB287721 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287721 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287721 Supplier Novus Biologicals Supplier No. NBP272876PE

Mouse Monoclonal Antibody

NARS2 Monoclonal antibody specifically detects NARS2 in Human samples. It is validated for Western Blot

Specifications

Antigen NARS2
Applications Western Blot
Classification Monoclonal
Clone OTI10C10
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias asnRS, asparagine tRNA ligase 2, mitochondrial (putative), Asparagine--tRNA ligase, asparaginyl-tRNA synthetase 2, mitochondrial (putative), EC 6.1.1, EC 6.1.1.22, FLJ23441, probable asparaginyl-tRNA synthetase, mitochondrial, SLM5
Host Species Mouse
Immunogen Full length human recombinant protein of human NARS2 (NP_078954) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79731
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.