Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAT2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
$191.80 - $437.00
Specifications
Antigen | NAT2 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145439
![]() |
Invitrogen™
PA5145439 |
200 μL |
Each for $437.00
|
|
|||||
FEPA5145440
|
Invitrogen™
PA5145440 |
20 μL |
Each for $191.80
|
|
|||||
Description
Participates in the detoxification of a plethora of hydrazine and arylamine drugs. Catalyzes the N- or O-acetylation of various arylamine and heterocyclic amine substrates and is able to bioactivate several known carcinogens.Specifications
NAT2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P11245, P50295, P50298 | |
10, 116632, 17961 | |
Recombinant Rat NAT2 protein, Asp20-Val280 (Accession #P50298). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
Aac2; Arylamide acetylase 2; arylamine N-acetyltransferase 2; AT-2; AV377607; N-acetyl transferase 2; N-acetyltransferase 2; N-acetyltransferase 2 (arylamine N-acetyltransferase); N-acetyltransferase type 2; N-Acetyltransferase-2 (arylamine N-acetyltransferase); Nat2; NAT-2; Nat2a; PNAT; Polymorphic arylamine N-acetyltransferase | |
NAT2 | |
IgG | |
Antigen affinity chromatography, Protein A | |
NAT2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title