Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAT2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | NAT2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
NAT2 Polyclonal specifically detects NAT2 in Human samples. It is validated for Western Blot.Specifications
| NAT2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 10 | |
| Synthetic peptides corresponding to NAT2(N-acetyltransferase 2 (arylamine N-acetyltransferase)) The peptide sequence was selected from the middle region of NAT2. Peptide sequence CLTEERGIWYLDQIRREQYITNKEFLNSHLLPKKKHQKIYLFTLEPRTIE. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| AAC2N-acetyltransferase type 2, Arylamide acetylase 2, arylamine N-acetyltransferase 2, EC 2.3.1.5, N-acetyltransferase 2 (arylamine N-acetyltransferase), NAT-2, PNAT, Polymorphic arylamine N-acetyltransferase | |
| NAT2 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title