Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAT8 Antibody (OTI5A8), Novus Biologicals™
SDP

Catalog No. NBP147863 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147863 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147863 Supplier Novus Biologicals Supplier No. NBP147863
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NAT8 Monoclonal specifically detects NAT8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen NAT8
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI5A8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50
Gene Alias Camello-like protein 1, CML1, EC 2.3.1, EC 2.3.1.-, GLAkidney- and liver-specific gene product, Hcml1, N-acetyltransferase 8, N-acetyltransferase 8 (GCN5-related, putative), probable N-acetyltransferase 8, TSC501N-acetyltransferase 8 (camello like), TSC510
Gene Symbols NAT8
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human NAT8 expression vector
Molecular Weight of Antigen 25.6 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9027
Test Specificity This antibody is specific for Homo sapiens N-acetyltransferase 8 (GCN5-related, putative) (NAT8).
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.