Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAT9 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$499.50

Specifications

Antigen NAT9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP153194
SDP
View Documents
Novus Biologicals
NBP153194
100 μL
Each of 1 for $499.50
Only null left
Add to Cart
 
Description

Description

NAT9 Polyclonal specifically detects NAT9 in Human samples. It is validated for Western Blot.
Specifications

Specifications

NAT9
Polyclonal
Rabbit
Q9BTE0
26151
Synthetic peptides corresponding to NAT9(N-acetyltransferase 9 (GCN5-related, putative)) Antibody(against the N terminal of NAT9. Peptide sequence MRLNQNTLLLGKKVVLVPYTSEHVPSRYHEWMKSEELQRLTASEPLTLEQ.
Primary
Western Blot
Unconjugated
RUO
DKFZP564C103, EBS, EBSP, hNATL, EC 2.3.1.-, embryo brain specific protein, Embryo brain-specific protein, N-acetyltransferase 9, N-acetyltransferase 9 (GCN5-related, putative)
NAT9
IgG
23 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.