Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Chemical Hazard Identification Labels

Catalog No. 17985467 Shop All National Marker Company Products
Click to view available options
Display Text:
Acetic Acid
Acetone
Chlorine
Diesel Fuel
Ethyl Alcohol
Flammable Liquid
Formaldehyde
Gasoline
Hydroxide Peroxide
Isopropyl Alcohol
Mercury
Methyl Alcohol
Nitric Acid
Phosphoric Acid
Propane
Toluene
Xylene

High visibility red-and-white pressure-sensitive vinyl labels ideal for alerting employees to potential chemical hazards in the workplace

  • Made to withstand most chemicals, solvents and vapors
  • Resist fading and scratching
  • For marking larger containers
  • Measure 6 × 10 in.

Specifications

Color White, Red
Format Label
Material Vinyl
Product Type Container Label
Display Text Phosphoric Acid
For Use With (Application) For marking larger containers, both indoor and outdoor applications
Quantity 1 Ea.
Width (English) 10 in.
Height (English) 6 in.
Height (Metric) 15.24 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.