Learn More
Thermo Scientific™ National Polypropylene and Glastic™ Vials with 8-425 Finish
Catalog No. p-4526449
Click to view available options
Volume (Metric) Working:
400 μL
Thermo Scientific™ National Polypropylene and Glastic Vials provide an economical alternative to glass for some applications.
Thermo Scientific™ National Polypropylene and Glastic Vials provide an economical alternative to glass for some applications.
  • Precision formed tips are designed to aid sample recovery for limited-volume applications
  • Glastic vial features a glass insert pre-inserted inside a clear TPX vial
TRUSTED_SUSTAINABILITY
Specifications
Description 600µL MicroVials
Color Clear
Material Polypropylene
Volume (Metric) 600 μL
Volume (Metric) Residual <4 μL
Volume (Metric) Working 400 μL
Closure Size 8-425
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.