Learn More
Thermo Scientific™ National Target™ Snap Cap Wide-Opening Vials
Catalog No. p-4526445
Click to view available options
Type:
Micro-V™ Amber Glass Snap-It Microsampling Vial
Micro-V™ Clear Glass Snap-It Microsampling Vial
Thermo Scientific™ National Target Snap Cap Wide-Opening Vials are easier to fill and provide a larger target area for the autosampler needle.
Thermo Scientific™ National Target Snap Cap Wide-Opening Vials are easier to fill and provide a larger target area for the autosampler needle.
  • 2mL, 12 x 32mm Crimp/Snap Ring Vials
  • Fit rotating or robotic arm autosamplers
  • I-D™ vials feature write-on patch with graduations at 0.5, 1.0 and 1.5mL
  • Target Snap-It™ vials can be used with snap caps or aluminum crimp seal closures
  • Fit most autosamplers that accommodate the 12 × 32mm vial format
  • Superior quality 33 expansion borosilicate clear glass (Type 1, Class A) or 51A amber glass
  • Uniformly flat bottom for security with inserts
TRUSTED_SUSTAINABILITY
Specifications
Description Micro-V Clear Glass Snap-It Microsampling Vial
Volume (Metric) 15 μL
Volume (Metric) Residual 15 μL
Type Micro-V™ Clear Glass Snap-It Microsampling Vial
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.