Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Native Lipoteichoic Acid Antibody (55), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2628110.025MG
Description
Native Lipoteichoic Acid Monoclonal specifically detects Native Lipoteichoic Acid in Bacterial samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunoassay.Specifications
| NativeLipoteichoicAcid | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.1% BSA with 0.02% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Store at 4degreesC. |
| Immunocytochemistry, Western Blot, Immunohistochemistry (Frozen), In situ PLA, Flow Cytometry, Immunofluorescence | |
| 55 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay | |
| LTA | |
| Microbial mixture of Streptococcus sobrims HG961, HG962, HG970 and HG977 | |
| 0.025 mg | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction