Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Native Lipoteichoic Acid Antibody (55), Novus Biologicals™

Mouse Monoclonal Antibody
$247.53 - $862.92
Specifications
| Antigen | NativeLipoteichoicAcid |
|---|---|
| Clone | 55 |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
Native Lipoteichoic Acid Monoclonal specifically detects Native Lipoteichoic Acid in Bacterial samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunoassay.Specifications
| NativeLipoteichoicAcid | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay | |
| Unconjugated | |
| Mouse | |
| PBS with 0.1% BSA with 0.02% Sodium Azide | |
| Microbial mixture of Streptococcus sobrims HG961, HG962, HG970 and HG977 | |
| Protein A purified |
| 55 | |
| Monoclonal | |
| Purified | |
| Bacteria | |
| LTA | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title