Learn More
Description
Specifications
Specifications
| Antigen | NativeLipoteichoicAcid |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), In Situ Hybridization (ISH), Western Blot |
| Classification | Monoclonal |
| Clone | 55 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay |
| Formulation | PBS with 0.1% BSA with 0.02% Sodium Azide |
| Gene Alias | LTA |
| Host Species | Mouse |
| Immunogen | Microbial mixture of Streptococcus sobrims HG961, HG962, HG970 and HG977 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
