Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Natriuretic Peptide Receptor C Antibody (1060830), HRP, Novus Biologicals™
SDP

Catalog No. NB305811 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305811 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305811 Supplier Novus Biologicals Supplier No. FAB102331H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Natriuretic Peptide Receptor C Monoclonal specifically detects Natriuretic Peptide Receptor C in Human samples. It is validated for Flow Cytometry, Western Blot.

Specifications

Antigen Natriuretic Peptide Receptor C
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1060830
Conjugate HRP
Dilution Western Blot, Flow Cytometry
Formulation PBS
Gene Alias ANP-C, ANPR-C, ANPRCFLJ14054, Atrial natriuretic peptide clearance receptor, atrial natriuretic peptide receptor 3, Atrial natriuretic peptide receptor type C, C5orf23, chromosome 5 open reading frame 23, GUCY2B, natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptidereceptor C), NPRC, NPR-C
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived human NPRC/NPR3, Met1-Glu481, Accession # P17342
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4883
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.