Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Natural Killer Cell Antibody (H25), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP2026A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP2026A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP2026A30 Supplier Novus Biologicals Supplier No. NBP250526AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Natural Killer Cell Monoclonal specifically detects Natural Killer Cell in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Natural Killer Cell
Applications Immunohistochemistry
Classification Monoclonal
Clone H25
Conjugate Alexa Fluor 350
Dilution Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias NK Cell
Host Species Mouse
Immunogen HSE-2 cell line
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.