Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Natural Killer Cell Antibody (H25), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309282 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309282 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309282 Supplier Novus Biologicals Supplier No. NBP250526MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Natural Killer Cell Monoclonal antibody specifically detects Natural Killer Cell in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Radioimmune Assays

Specifications

Antigen Natural Killer Cell
Applications Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone H25
Conjugate mFluor Violet 450 SE
Dilution Immunohistochemistry, Immunoprecipitation, Radioimmunoassay
Formulation 50mM Sodium Borate
Gene Alias NK Cell
Host Species Mouse
Immunogen HSE-2 cell line
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.