Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nav1.5 Antibody, Novus Biologicals™
SDP

Catalog No. NB600804 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600804 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600804 Supplier Novus Biologicals Supplier No. NB600804
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Nav1.5 Polyclonal specifically detects Nav1.5 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen Nav1.5
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 8 - 10 μg/mL, Immunohistochemistry-Paraffin 8 - 10 μg/mL, Peptide ELISA Detection limit 1:2000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_932173.1
Gene Alias cardiac sodium channel alpha subunit, cardiac tetrodotoxin-insensitive voltage-dependent sodium channel alpha subunit, CDCD2VF1, HB1sodium channel, voltage-gated, type V, alpha (long QT syndrome 3), HB2, HBBD, HH1CMD1E, ICCD, IVF, LQT3SSS1CMPD2, Nav1.5, PFHB1, Sodium channel protein cardiac muscle subunit alpha, sodium channel protein type 5 subunit alpha, sodium channel protein type V alpha subunit, Sodium channel protein type V subunit alpha, sodium channel, voltage-gated, type V, alpha subunit, Voltage-gated sodium channel subunit alpha Nav1.5
Gene Symbols SCN5A
Host Species Goat
Immunogen Peptide with sequence C-ETDDQSPEKIN, corresponding to the internal region according to NP_932173.1; NP_000326.2; NP_001092874.1; NP_001092875.1; NP_001153632.1; NP_001153633.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6331
Test Specificity This antibody is expected to recognize all reported isoforms (NP_932173.1; NP_000326.2; NP_001092874.1; NP_001092875.1).
Target Species Human, Mouse, Rat, Bovine, Canine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.