Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nav1.7 Antibody (N68/6), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP212904Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212904Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212904Z Supplier Novus Biologicals Supplier No. NBP212904AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nav1.7 Monoclonal specifically detects Nav1.7 in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Nav1.7
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone S68-6
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Alias ETHA, FEB3B, NE-NA, NENAVoltage-gated sodium channel subunit alpha Nav1.7, Neuroendocrine sodium channel, Peripheral sodium channel 1, PN1hNE-Na, sodium channel protein type 9 subunit alpha, Sodium channel protein type IX subunit alpha, sodium channel, voltage-gated, type IX, alpha subunit, voltage-gated sodium channel alpha subunit Nav1.7, voltage-gated, type IX, alpha polypeptide
Gene Symbols SCN9A
Host Species Mouse
Immunogen Fusion protein amino acids 1751-1946 (C-terminus) of human Nav1.7
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6335
Test Specificity Detects approx 230kDa. No cross-reactivity against other Nav channels.
Target Species Human, Mouse, Rat, Hamster
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.