Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nav1.8 Antibody (S134), Novus Biologicals™
SDP

Catalog No. NBP147615 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147615 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147615 Supplier Novus Biologicals Supplier No. NBP147615
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nav1.8 Monoclonal specifically detects Nav1.8 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Microarray.

Specifications

Antigen Nav1.8
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S134-12
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:1000
Gene Accession No. Q62968
Gene Alias hPN3, Nav1.8, PN3, sodium channel protein type 10 subunit alpha, sodium channel protein type X subunit alpha, sodium channel, voltage-gated, type X, alpha subunit, voltage-gated, type X, alpha polypeptide
Gene Symbols SCN10A
Host Species Mouse
Immunogen Fusion protein amino acids 1724-1956 of rat Nav1.8
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6336
Test Specificity Detects approx 220kDa. No cross reactivity against other Nav channels.
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.