Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAV1 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB135389 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB135389 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB135389 Supplier Novus Biologicals Supplier No. NBP306303FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NAV1 Polyclonal specifically detects NAV1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen NAV1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias FLJ12560, FLJ14203, KIAA1151POMFIL3DKFZp781D0314, KIAA1213, MGC14961, neuron navigator 1, neuron navigator-1, pore membrane and/or filament interacting like protein 3, Pore membrane and/or filament-interacting-like protein 3, Steerin-1, STEERIN1, Unc-53 homolog 1, unc53H1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NAV1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89796
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.