Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAV3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB170116 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170116 25 μg
NB170115 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB170116 Supplier Novus Biologicals Supplier No. NBP31752025UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NAV3 Polyclonal antibody specifically detects NAV3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen NAV3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias KIAA0938pore membrane and/or filament interacting like protein 1, neuron navigator 3, POMFIl1, POMFIL1steerin 3, Pore membrane and/or filament-interacting-like protein 1, steerin-3, STEERIN3, Unc-53 homolog 3, unc53H3
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: SPTFRRLFGAKAGGKSASAPNTEGVKSSSVMPSPSTTLARQGSLESPSSGTGSMGSAGGLSGSSSPLFNKPSD
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 89795
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.