Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ NBC 6

Catalog No. p-7199106
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

NLRP3 inflammasome inhibitor

NBC 6 is a NLRP3 inflammasome inhibitor (IC50 = 574 nM). Inhibits nigericin-induced inflammasome activation in THP-1 cells and Imiquimod (Cat. No. 3700) induced IL-1β release from LPS-primed bone marrow-derived macrophages (BMDMs).

Specifications

Chemical Name or Material (T4)-[2-Acetyl-4,4,4-trichloro-3-(imino-κN)butanamidito-κO]diphenylboron
CAS 2068818-02-8
Quantity 10 mg
Target Inflammasomes Inhibitors
Molecular Formula C18H16BCl3N2O2
Formula Weight 1869
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.