Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NBEAL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | NBEAL1 |
---|---|
Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
NBEAL1 Polyclonal specifically detects NBEAL1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NBEAL1 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
A530083I02Rik, ALS2CR16, ALS2CR17FLJ22838, amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 16, amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 17, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 16 protein, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 17 protein, beach, FLJ26555, MGC164581, MGC168834, neurobeachin-like 1, neurobeachin-like protein 1 | |
NBEAL1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
Q6ZS30 | |
65065 | |
This antibody was developed against a recombinant protein corresponding to amino acids: QKDPDYLKLWLDTFVSSYEQFLDVDFEKLPTRVDDMPPGISLLPDNILQVLRIQLLQCVQKMADGLEE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title