Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NBI 35965 hydrochloride
GREENER_CHOICE

Catalog No. 310010
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective CRF1 antagonist

Potent and selective corticotropin-releasing factor receptor 1 (CRF1) antagonist (Ki values are 4 and > 10000 nM at CRF1 and CRF2 respectively). Inhibits cAMP accumulation and adrenocorticotropic hormone (ACTH) production in vitro (pIC50 values are 7.1 and 6.9 respectively) and attenuates CRF and ACTH production in vivo. Orally active and brain penetrant with anxiolytic activity.

Chemical Identifiers

CAS 603151-83-3
Molecular Formula C21H23Cl3N4

Specifications

Quantity 10 mg
Formula Weight 437.79
Purity >98%
Molecular Weight (g/mol) 437.793
Chemical Name or Material NBI 35965 hydrochloride
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.