Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ NBOH-2C-CN hydrochloride
Click to view available options
Quantity:
10 mg
50 mg
Description
High affinity, selective 5-HT2A agonist (Ki = 1.3 nM). Exhibits 100-fold selectivity for 5-HT2A over 5-HT2C. Elicits head twitch behavior and reduces anxiety-related marble-burying activity in mice.
Specifications
Specifications
Quantity | 10 mg |
Product Type | NBOH-2C-CN hydrochloride |
Purity | >98% |
Molecular Weight (g/mol) | 348.82 |
Inhibitors | NBOH-2C-CN hydrochloride |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction