Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NBPF15 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB133153 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB133153 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB133153 Supplier Novus Biologicals Supplier No. NBP306172G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NBPF15 Polyclonal specifically detects NBPF15 in Human, Cynomolgus Monkey samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen NBPF15
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias AB14, Ag3, NBPF16, Neuroblastoma Breakpoint Family Member 15, Neuroblastoma Breakpoint Family Member 16, Neuroblastoma Breakpoint Family, Member 15, Neuroblastoma Breakpoint Family, Member 16
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NBPF15 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284565
Target Species Human, Cynomolgus Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.