Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NBR1 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP171703UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP171703UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP171703UV Supplier Novus Biologicals Supplier No. NBP171703UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NBR1 Polyclonal specifically detects NBR1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen NBR1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Cell migration-inducing gene 19 protein, FLJ98272, KIAA0049CA125, M17S2FLJ55359,1A1-3B, Membrane component chromosome 17 surface marker 2,1A13B, membrane component, chromosome 17, surface marker 2 (ovarian carcinoma antigenCA125), migration-inducing protein 19, neighbor of BRCA1 gene 1, Neighbor of BRCA1 gene 1 protein, next to BRCA1 gene 1 protein, Protein 1A1-3B
Gene Symbols NBR1
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human NBR1 protein (within residues 50-250). [Swiss-Prot Q14596]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4077
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.