Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nbs1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NB100143

 View more versions of this product

Catalog No. NB100143


Only null left
Add to Cart

Description

Description

Nbs1 Polyclonal specifically detects Nbs1 in Human, Mouse, Hamster, Mammal, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, In-situ Hybridization, SDS-Page, Knockout Validated.
Specifications

Specifications

Nbs1
Polyclonal
Western Blot 1:1000. Use reported in multiple pieces of scientific literature, Chromatin Immunoprecipitation reported in multiple pieces of scientific literature, Flow Cytometry reported in multiple pieces of scientific literature, ELISA, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200. Use reported in multiple pieces of scientific literature, Immunoprecipitation 3 ul. Use reported in multiple pieces of scientific literature, Immunohistochemistry-Paraffin 1:100 - 1:200, Immunoblotting, In-situ Hybridization reported in scientific literature (PMID 26415217), SDS-Page, Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated
O60934
NBN
Nbs1 Antibody was made to a partial length human NBS1 protein [Swiss-Prot: O60934].
Unpurified
RUO
Primary
Human, mouse, hamster and monkey. Mammal reactivity reported in scientific literature (PMID: 24064949).
Store at 4C. Do not freeze.
IgG
Western Blot, ChIP Assay, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, In Situ Hybridization (ISH), Immunoassay
Unconjugated
Whole antisera with 0.02% Sodium Azide
ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex
Rabbit
85 kDa
0.05 mL
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Tumor Suppressors
4683
Human, Mouse, Hamster, Mammalia, Primate
Antisera
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only