Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nbs1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$354.50 - $581.50

Specifications

Antigen Nbs1
Dilution Western Blot 1:1000. Use reported in multiple pieces of scientific literature, Chromatin Immunoprecipitation reported in multiple pieces of scientific literature, Flow Cytometry reported in multiple pieces of scientific literature, ELISA, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200. Use reported in multiple pieces of scientific literature, Immunoprecipitation 3 ul. Use reported in multiple pieces of scientific literature, Immunohistochemistry-Paraffin 1:100 - 1:200, Immunoblotting, In-situ Hybridization reported in scientific literature (PMID 26415217), SDS-Page, Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated
Applications Western Blot, ChIP Assay, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, In Situ Hybridization (ISH), Immunoassay
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB100143SS
SDP
View Documents
Novus Biologicals
NB100143SS
0.025 mL
Each for $354.50
Only null left
Add to Cart
 
NB100143
SDP
View Documents
Novus Biologicals
NB100143
0.05 mL
Each for $581.50
Only null left
Add to Cart
 
Description

Description

Nbs1 Polyclonal specifically detects Nbs1 in Human, Mouse, Hamster, Mammal, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, In-situ Hybridization, SDS-Page, Knockout Validated.
Specifications

Specifications

Nbs1
Western Blot, ChIP Assay, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, In Situ Hybridization (ISH), Immunoassay
Unconjugated
Rabbit
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Tumor Suppressors
Whole antisera with 0.02% Sodium Azide
ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex
NBN
IgG
Unpurified
Human, mouse, hamster and monkey. Mammal reactivity reported in scientific literature (PMID: 24064949).
Western Blot 1:1000. Use reported in multiple pieces of scientific literature, Chromatin Immunoprecipitation reported in multiple pieces of scientific literature, Flow Cytometry reported in multiple pieces of scientific literature, ELISA, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200. Use reported in multiple pieces of scientific literature, Immunoprecipitation 3 ul. Use reported in multiple pieces of scientific literature, Immunohistochemistry-Paraffin 1:100 - 1:200, Immunoblotting, In-situ Hybridization reported in scientific literature (PMID 26415217), SDS-Page, Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated
Polyclonal
Antisera
RUO
Human, Mouse, Hamster, Mammalia, Primate
O60934
4683
Nbs1 Antibody was made to a partial length human NBS1 protein [Swiss-Prot: O60934].
Primary
Store at 4C. Do not freeze.
85 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.