Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nbs1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100143SS
Description
Nbs1 Polyclonal specifically detects Nbs1 in Human, Mouse, Hamster, Mammal, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, In-situ Hybridization, SDS-Page, Knockout Validated.Specifications
Nbs1 | |
Polyclonal | |
Western Blot 1:1000. Use reported in multiple pieces of scientific literature, Chromatin Immunoprecipitation reported in multiple pieces of scientific literature, Flow Cytometry reported in multiple pieces of scientific literature, ELISA, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200. Use reported in multiple pieces of scientific literature, Immunoprecipitation 3 ul. Use reported in multiple pieces of scientific literature, Immunohistochemistry-Paraffin 1:100 - 1:200, Immunoblotting, In-situ Hybridization reported in scientific literature (PMID 26415217), SDS-Page, Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated | |
O60934 | |
NBN | |
Nbs1 Antibody was made to a partial length human NBS1 protein [Swiss-Prot: O60934]. | |
Unpurified | |
RUO | |
Primary | |
Human, mouse, hamster and monkey. Mammal reactivity reported in scientific literature (PMID: 24064949). | |
Store at 4C. Do not freeze. | |
IgG |
Western Blot, ChIP Assay, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, In Situ Hybridization (ISH), Immunoassay | |
Unconjugated | |
Whole antisera with 0.02% Sodium Azide | |
ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex | |
Rabbit | |
85 kDa | |
0.025 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Tumor Suppressors | |
4683 | |
Human, Mouse, Hamster, Mammalia, Primate | |
Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction