Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nbs1 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP106609G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106609G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106609G Supplier Novus Biologicals Supplier No. NBP106609G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nbs1 Polyclonal specifically detects Nbs1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Nbs1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex
Gene Symbols NBN
Host Species Rabbit
Immunogen Nbs1 Antibody was made to a synthetic peptide made to an internal portion of the mouse NBS1 protein (within residues 350-400). [Swiss-Prot# Q9R207]
Molecular Weight of Antigen 97 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4683
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.