Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nbs1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10060648 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10060648 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10060648 Supplier Novus Biologicals Supplier No. NB10060648
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nbs1 Polyclonal specifically detects Nbs1 in Mouse samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen Nbs1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q9R207
Gene Alias ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex
Gene Symbols NBN
Host Species Rabbit
Immunogen The immunogen recognized by Nbs1 Antibody maps to a region between residue 701 and 751 of mouse Nibrin using the numbering given in entry NP_038780.3 (GeneID 27354).
Molecular Weight of Antigen 84 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4683
Test Specificity Mouse. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Dog, Horse, Guinea pig_10141, Panda, White-tufted-ear marmoset, Naked mole rat, African elephant, Little brown bat, Common squirrel monkey, and Thirteen-lined ground squirrel.
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.