Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nbs1 Rabbit anti-Human, Clone: 4G4R1, Novus Biologicals™
SDP

Catalog No. NB167801 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167801 100 μg
NB167802 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167801 Supplier Novus Biologicals Supplier No. NBP316355100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Nbs1 Monoclonal antibody specifically detects Nbs1 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen Nbs1
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 4G4R1
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 655-754 of human Nbs1 (O60934).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4683
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.