Learn More
Description
Specifications
Specifications
| Antigen | NCAM-1/CD56 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 123C3.D5 |
| Conjugate | HRP |
| Dilution | Western Blot 1:100-1:2000, ELISA 1-5 μg/mL for coating, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Gene Alias | CD56, CD56 NCAM-1, CD56 antigen, MSK39, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM |
| Gene Symbols | NCAM1 |
| Host Species | Mouse |
| Immunogen | Membrane preparation of a small cell lung carcinoma. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
