Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCAM-1/CD56 Antibody (2524C), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB313484 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313484 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313484 Supplier Novus Biologicals Supplier No. FAB24086APCCY7
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NCAM-1/CD56 Monoclonal specifically detects NCAM-1/CD56 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen NCAM-1/CD56
Applications Flow Cytometry
Classification Monoclonal
Clone 2524C
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD56, CD56/ NCAM-1, CD56 antigen, MSK39, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM
Host Species Rabbit
Immunogen Mouse myeloma cell line, NS0-derived human NCAM -1/CD56, Leu20-Pro603 and Glu636-Asn741, Accession # NP_001070150
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Membrane Vesicle Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4684
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.