Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCAM-1/CD56 Antibody (735), Alexa Fluor™ 594 - Chimeric, Novus Biologicals™
SDP

Catalog No. NP22669A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP22669A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP22669A94 Supplier Novus Biologicals Supplier No. NBP252669AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NCAM-1/CD56 Monoclonal specifically detects NCAM-1/CD56 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen NCAM-1/CD56
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 735
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD56, CD56/ NCAM-1, CD56 antigen, MSK39, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM
Gene Symbols NCAM1
Host Species Rabbit
Immunogen Homopolymers of alpha 2,8-linked sialic acid of Escherichia coli K1.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4684
Test Specificity Recognises Polysialic acid (PSA), a carbohydrate polymer attached to the neural cell adhesion molecule (NCAM), which shares molecular epitopes with homopolymers of (alpha)2,8-linked sialic acid of Escherichia coli K1.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.