Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCAM-1/CD56 Antibody (RNL-1), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP197717V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197717V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197717V Supplier Novus Biologicals Supplier No. NBP197717V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NCAM-1/CD56 Monoclonal specifically detects NCAM-1/CD56 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen NCAM-1/CD56
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone RNL-1
Conjugate DyLight 405
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Alias CD56, CD56 NCAM-1, CD56 antigen, MSK39, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM
Gene Symbols NCAM1
Host Species Mouse
Immunogen Clone RNL-1 is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with the small cell lung cancer cell line NCI-H82.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Astrocyte Markers, Cellular Markers, Cytokine Research, Cytoskeleton Markers, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Membrane Vesicle Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4684
Test Specificity RNL-1 was defined as a cluster I antibody during the Second International Workshop on Small Cell Lung Cancer (SCLC) Antibodies. RNL-1 recognizes NCAM which is present in small cell lung cancer (SCLC) and lung carcinoids.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.