Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCAPD3 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001573 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001573 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001573 Supplier Novus Biologicals Supplier No. NB1001573
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

NCAPD3 Polyclonal specifically detects NCAPD3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot.

Specifications

Antigen NCAPD3
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 2-10 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. P42695
Gene Alias CAPD3, CAP-D3, FLJ42888, hCAP-D3condensin-2 complex subunit D3, hcp-6, hHCP-6, KIAA0056MGC104671, Non-SMC condensin II complex subunit D3, non-SMC condensin II complex, subunit D3
Gene Symbols NCAPD3
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1450 and the C-terminus (residue 1507) of human Chromosome-Associated Protein D3 using the numbering given in entry NP_056076.1 (GeneID 23310).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 23310
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Gorilla, Chimpanzee, Crab-eating macaque and Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.