Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCBP1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP241222 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP241222 0.1 mg
NB435948 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP241222 Supplier Novus Biologicals Supplier No. NBP241222
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NCBP1 Polyclonal specifically detects NCBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NCBP1
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.02% Sodium Azide
Gene Alias CBP80nuclear cap binding protein subunit 1, 80kD, NCBP 80 kDa subunit, NCBPMGC2087, nuclear cap binding protein subunit 1, 80kDa, nuclear cap-binding protein subunit 1,80 kDa nuclear cap-binding protein, Sto1
Gene Symbols NCBP1
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid peptide near the center of human NCBP1. The immunogen is located within amino acids 420 - 470 of NCBP1. Amino Acid Squence: LSQDPESPKPKFVREV
Molecular Weight of Antigen 87 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4686
Test Specificity At least two isoforms of NCBP1 are known to exist; this antibody will detect both isoforms of NCBP1.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.