Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583370
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA583370 100 μL
1 options

Catalog No. PIPA583370

Supplier: Invitrogen™ PA583370

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AYNMVPPKVK CLTKIWHPNI TETGEICLSL LREHSIDGTG WAPTRTLKDV VWGLNSLFTD LLNFDDPLNI EAAEHHLRDK EDFRNKVDDY Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 98%.

Accepts the ubiquitin-like protein NEDD8 from the UBA3-NAE1 E1 complex and catalyzes its covalent attachment to other proteins. The specific interaction with the E3 ubiquitin ligase RBX2, but not RBX1, suggests that the RBX2-UBE2F complex neddylates specific target proteins, such as CUL5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCE2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBE2F
Gene Accession No. Q969M7
Gene Alias 2510010F15Rik; AI851109; MGC18120; NCE2; NEDD8 carrier protein UBE2F; NEDD8 protein ligase UBE2F; NEDD8-conjugating enzyme 2; NEDD8-conjugating enzyme UBE2F; RGD1307608; RING-type E3 NEDD8 transferase UBE2F; UBE2F; Ube2fa; ubiquitin conjugating enzyme E2 F (putative); ubiquitin conjugating enzyme E2F (putative); ubiquitin-conjugating enzyme E2 F; ubiquitin-conjugating enzyme E2F (putative)
Gene Symbols UBE2F
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBE2F. Recombinant protein control fragment (Product #RP-96214).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140739
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.