Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCF4 Antibody (OTI4B5), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP272893G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272893G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272893G Supplier Novus Biologicals Supplier No. NBP272893G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NCF4 Monoclonal specifically detects NCF4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NCF4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4B5
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias MGC3810, NCF, NCF-4, neutrophil cytosol factor 4, neutrophil cytosolic factor 4 (40kD), neutrophil cytosolic factor 4, 40kDa, Neutrophil NADPH oxidase factor 4, p40phox, p40-phox, SH3 and PX domain-containing protein 4, SH3PXD4P40PHOX
Gene Symbols NCF4
Host Species Mouse
Immunogen Full length recombinant protein of human NCF4 (NP_000622) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4689.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.