Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCKAP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP226086 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226086 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP226086 Supplier Novus Biologicals Supplier No. NBP226086
Add to Cart
Add to Cart

Goat Polyclonal Antibody

NCKAP1 Polyclonal specifically detects NCKAP1 in Human, Mouse, Rat, Porcine, Bovine, Canine samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen NCKAP1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias FLJ11291, HEM2p125Nap1, KIAA0587, Membrane-associated protein HEM-2, MGC8981, NAP 1, NAP125, NAP1Nap1, NCK-associated protein 1
Gene Symbols NCKAP1
Host Species Goat
Immunogen Peptide with sequence C-TPREYLTSHLEIR corresponding to internal region according to NP_995314.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 10787
Test Specificity This antibody is expected to recognize both reported isoforms (NP_038464.1; NP_995314.1).
Target Species Human, Mouse, Rat, Pig, Bovine, Canine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.