Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCKAP5L Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB002995 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002995 Supplier Novus Biologicals Supplier No. NBP278830

Rabbit Polyclonal Antibody

NCKAP5L Polyclonal specifically detects NCKAP5L in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen NCKAP5L
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.04 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunoprecipitation 50-100 μL/mg lysate
Formulation Tris-buffered Saline, 0.1% BSA with 0.09% Sodium Azide
Gene Alias FLJ36581, KIAA1602, NCK-associated protein 5-like
Gene Symbols NCKAP5L
Host Species Rabbit
Immunogen The epitope recognized by A305-888A-M maps to a region between residue 1025 and 1075 of human Nck-associated protein 5-like using the numbering given in entry Q9HCH0.2 (GeneID 57701).
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 57701
Target Species Human, Mouse
Content And Storage Store at 2-8°C/ 1 year from date of receipt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.