Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCKIPSD Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB329491 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329491 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329491 Supplier Novus Biologicals Supplier No. NBP300167APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NCKIPSD Polyclonal antibody specifically detects NCKIPSD in Human samples. It is validated for ELISA

Specifications

Antigen NCKIPSD
Applications ELISA
Classification Polyclonal
Conjugate APC
Dilution ELISA
Formulation PBS
Gene Alias 54 kDa vimentin-interacting protein, 90 kDa SH3 protein interacting with Nck, AF3p21, AF3P21DIP-1, dia interacting protein, Dia-interacting protein 1, diaphanous protein interacting protein, Diaphanous protein-interacting protein, DIP, DIP1, MGC23891, NCK interacting protein with SH3 domain, ORF1, SPIN90VIP54, WASLBP, WASP-interacting SH3-domain protein, WISHSH3 adapter protein SPIN90, Wisk90 kDa
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human NCKIPSD (Uniprot#: Q9NZQ3-3; Met1-Thr244)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51517
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.