Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCOA2 Antibody (OTI3C7), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB287711 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287711 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287711 Supplier Novus Biologicals Supplier No. NBP272894APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NCOA2 Monoclonal antibody specifically detects NCOA2 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen NCOA2
Applications Western Blot
Classification Monoclonal
Clone OTI3C7
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias BHLHE75, bHLHe75MGC138808, Class E basic helix-loop-helix protein 75, EC 2.3.1.48, GRIP1glucocorticoid receptor-interacting protein-1, KAT13C, NCoA-2SRC2, nuclear receptor coactivator 2, TIF2hTIF2, Transcriptional intermediary factor 2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-280 of human NCOA2 (NP_006531) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 10499
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.