Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCOA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB093143 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093143 100 μL
NB093144 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093143 Supplier Novus Biologicals Supplier No. NBP303763100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NCOA2 Polyclonal specifically detects NCOA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.

Specifications

Antigen NCOA2
Applications Western Blot, Immunohistochemistry, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias BHLHE75, bHLHe75MGC138808, Class E basic helix-loop-helix protein 75, EC 2.3.1.48, GRIP1glucocorticoid receptor-interacting protein-1, KAT13C, NCoA-2SRC2, nuclear receptor coactivator 2, TIF2hTIF2, Transcriptional intermediary factor 2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 550-820 of human NCOA2 (NP_006531.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 10499
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.