Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCOA3/AIB1 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB292189 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292189 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292189 Supplier Novus Biologicals Supplier No. NB100314F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NCOA3/AIB1 Polyclonal antibody specifically detects NCOA3/AIB1 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen NCOA3/AIB1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias ACTRCBP-interacting protein, AIB1AIB-1, Amplified in breast cancer 1 protein, BHLHE42, bHLHe42p/CIP, CAGH16, Class E basic helix-loop-helix protein 42, CTG26, KAT13B, MGC141848, NCoA-3, nuclear receptor coactivator 3, pCIP, RAC-3, RAC3pCIP, Receptor-associated coactivator 3, SRC3, SRC-3EC 2.3.1.48, Steroid receptor coactivator protein 3, Thyroid hormone receptor activator molecule 1, TNRC16, TRAM1, TRAM-1TNRC14
Host Species Rabbit
Immunogen A synthetic peptide, which represented a portion of human nuclear receptor coactivator 3 encoded within exon 13 (LocusLink ID 8202).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Chromatin Research, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 8202
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.