Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCOA3/AIB1 Antibody, Novus Biologicals™
SDP

Catalog No. NB300567 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300567 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300567 Supplier Novus Biologicals Supplier No. NB300567

Rabbit Polyclonal Antibody

NCOA3/AIB1 Polyclonal specifically detects NCOA3/AIB1 in Human samples. It is validated for Western Blot.

Specifications

Antigen NCOA3/AIB1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml
Formulation PBS, 1 mg/ml BSA with 0.05% Sodium Azide
Gene Alias ACTRCBP-interacting protein, AIB1AIB-1, Amplified in breast cancer 1 protein, BHLHE42, bHLHe42p/CIP, CAGH16, Class E basic helix-loop-helix protein 42, CTG26, KAT13B, MGC141848, NCoA-3, nuclear receptor coactivator 3, pCIP RAC-3, RAC3pCIP, Receptor-associated coactivator 3, SRC3, SRC-3EC 2.3.1.48, Steroid receptor coactivator protein 3, Thyroid hormone receptor activator molecule 1, TNRC16, TRAM1, TRAM-1TNRC14
Gene Symbols NCOA3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues G(3) L G E N L D P L A S D S(15) C of human AIB1.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 8202
Test Specificity Detects amplified in breast cancer 1 (nuclear receptor coactivator 3) from human tissues and cells.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.