Learn More
Description
Specifications
Specifications
| Antigen | NCOA3/AIB1 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.0mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 |
| Formulation | 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal |
| Gene Alias | ACTRCBP-interacting protein, AIB1AIB-1, Amplified in breast cancer 1 protein, BHLHE42, bHLHe42p/CIP, CAGH16, Class E basic helix-loop-helix protein 42, CTG26, KAT13B, MGC141848, NCoA-3, nuclear receptor coactivator 3, pCIP RAC-3, RAC3pCIP, Receptor-associated coactivator 3, SRC3, SRC-3EC 2.3.1.48, Steroid receptor coactivator protein 3, Thyroid hormone receptor activator molecule 1, TNRC16, TRAM1, TRAM-1TNRC14 |
| Gene Symbols | NCOA3 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
