Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCOA4 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB160382 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160382 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB160382 Supplier Novus Biologicals Supplier No. NBP318136
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NCOA4 Polyclonal antibody specifically detects NCOA4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen NCOA4
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry 1 ug/ml, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Androgen receptor coactivator 70 kDa protein, Androgen receptor-associated protein of 70 kDa, ARA70RFGret fused, DKFZp762E1112, ELE1NCoA-4, nuclear receptor coactivator 4,70 kDa AR-activator, PTC3RET-activating gene ELE1, Ret-activating protein ELE1,70 kDa androgen receptor coactivator
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 13 amino acids near the center of human NCOA4. The immunogen is located within 420-470 amino acids of NCOA4.
Purification Method Affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8031
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.